3-Isobutoxy-5-(trifluoromethoxy)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-6376-5G
BB-6376-1G
COBBBB-6376-5GEA
0
GBP
COBBBB-6376-5G
COBBBB-6376-1G
3-Isobutoxy-5-(trifluoromethoxy)phenylboronic acid
3-Isobutoxy-5-(trifluoromethoxy)phenylboronic acid
Formula:
C₁₁H₁₄BF₃O₄ |
MDL Number:
MFCD16295064 CAS Number: 1256346-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...