(4-Chlorophenylthio)acetone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-((4-Chlorophenyl)thio)propan-2-one, 1-(4-Chlorophenylthio)propan-2-one
M-Clarity™ quality level = MQ100
653594-25G
653594-5G
SIAL653594-25GEA
210
GBP
SIAL653594-25G
SIAL653594-5G
(4-Chlorophenylthio)acetone, Sigma-Aldrich®
(4-Chlorophenylthio)acetone
Formula:
C₉H₉ClOS MW: 200.6852 g/mol |
MDL Number:
MFCD00013644 CAS Number: 25784-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...