4-Bromo-2,6-dinitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23899G1 AS23899G5
ACTIAS23899G1EA 234 GBP
ACTIAS23899G1 ACTIAS23899G5
4-Bromo-2,6-dinitrobenzoic acid
4-Bromo-2,6-dinitrobenzoic acid
Formula: C₇H₃BrN₂O₆
MDL Number: MFCD06410771
CAS Number: 95192-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR