(E)-1-Hexene-1,2-diboronic acid bis(pinacol)ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010764-2G 010764-5G
FLUO010764-2GEA 0 GBP
FLUO010764-2G FLUO010764-5G
(E)-1-Hexene-1,2-diboronic acid bis(pinacol)ester
(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester
Formula: C₁₈H₃₄B₂O₄
MW: 336.09 g/mol
MDL Number: MFCD03093898
CAS Number: 185427-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR