(E)-1-Heptene-1,2-diboronic acid bis(pinacol)ester ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L19649.03 L19649.06
L19649.03EA 57.8 GBP
L19649.03 L19649.06
(E)-1-Heptene-1,2-diboronic acid bis(pinacol)ester ≥98%
(E)-1-Heptene-1,2-diboronic acid bis(pinacol) ester

Purity: 98%
CAS No.: 307531-74-4
Molecular Formula: C19H36B2O4
Formula Weight: 350.12
Flash Point: >110°(230°F)
Refractive Index: 1.4560
Boiling Point: 108°/0.08mm
Beilstein Reference: 8859280
MDL No.: MFCD02683463

Formula: C₁₉H₃₆B₂O₄
MDL Number: MFCD02683463
CAS Number: 307531-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR