Methyl-1,2,4-triazole-3-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate

AS24432G100 AS24432G25
ACTIAS24432G100EA 0 GBP
ACTIAS24432G100 ACTIAS24432G25
Methyl-1,2,4-triazole-3-carboxylate
Methyl 1,2,4-triazole-3-carboxylate
Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
MDL Number: MFCD00135989
CAS Number: 4928-88-5
EINECS: 480-470-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR