1-(2-Cyanophenyl)piperidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(1-Piperidinyl)benzonitrile, 2-Piperidinobenzonitrile

CFF300.5 017342-5G
CHMPCFF300.5EA 0 GBP
CHMPCFF300.5 FLUO017342-5G
1-(2-Cyanophenyl)piperidine
1-(2-Cyanophenyl)piperidine
Formula: C₁₂H₁₄N₂
MW: 186.26 g/mol
MDL Number: MFCD00049221
CAS Number: 72752-52-4
EINECS: 427-330-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR