1-(2-(4-Nitrophenoxy)ethyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1216-5G AN-1216-25G AN-1216-1G
COBBAN-1216-5GEA 0 GBP
COBBAN-1216-5G COBBAN-1216-25G COBBAN-1216-1G
1-(2-(4-Nitrophenoxy)ethyl)piperidine
1-(2-(4-Nitrophenoxy)ethyl)piperidine
Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD01308502
CAS Number: 92033-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR