1-Boc-4-(4-aminophenoxy)piperidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
tert-Butyl 4-(4-aminophenoxy)piperidine-1-carboxylate, 4-{[1-(tert-Butoxycarbonyl)piperidin-4-yl]oxy}aniline
AS12887G1
AS12887G5
ACTIAS12887G1EA
136
GBP
ACTIAS12887G1
ACTIAS12887G5
1-Boc-4-(4-aminophenoxy)piperidine
1-Boc-4-(4-aminophenoxy)piperidine
Formula:
C₁₆H₂₄N₂O₃ MW: 292.3734 g/mol |
MDL Number:
MFCD04115023 CAS Number: 138227-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...