4-(3-Chlorophenoxy)piperidine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2058-1G HC-2058-5G
COBBHC-2058-1GEA 0 GBP
COBBHC-2058-1G COBBHC-2058-5G
4-(3-Chlorophenoxy)piperidine hydrochloride
4-(3-Chlorophenoxy)piperidine hydrochloride
Formula: C₁₁H₁₅Cl₂NO
MDL Number: MFCD03840130
CAS Number: 65367-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR