1-Boc-4-(4-bromophenoxy)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Boc-4-(4-Bromophenoxy)piperidine

HC-2010-5G HC-2010-1G
COBBHC-2010-5GEA 0 GBP
COBBHC-2010-5G COBBHC-2010-1G
1-Boc-4-(4-bromophenoxy)piperidine
1-Boc-4-(4-bromophenoxy)piperidine
Formula: C₁₆H₂₂BrNO₃
MW: 356.2548 g/mol
MDL Number: MFCD11053889
CAS Number: 308386-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR