1-Boc-4-(4-bromo-3,5-dimethylphenoxy)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2134-5G HC-2134-1G
COBBHC-2134-5GEA 0 GBP
COBBHC-2134-5G COBBHC-2134-1G
1-Boc-4-(4-bromo-3,5-dimethylphenoxy)piperidine
1-Boc-4-(4-bromo-3,5-dimethylphenoxy)piperidine
Formula: C₁₈H₂₆BrNO₃
MDL Number: MFCD11504904
CAS Number: 1072944-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR