1-Boc-4-(2-bromo-4-nitrophenoxy)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2114-1G HC-2114-5G
COBBHC-2114-1GEA 0 GBP
COBBHC-2114-1G COBBHC-2114-5G
1-Boc-4-(2-bromo-4-nitrophenoxy)piperidine
1-Boc-4-(2-bromo-4-nitrophenoxy)piperidine
Formula: C₁₆H₂₁BrN₂O₅
MDL Number: MFCD11504894
CAS Number: 337520-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR