5-(3-Pyridyl)-1H-tetrazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(2H-Tetrazol-5-yl)pyridine

CFF679.1 005512-1G 005512-5G
CHMPCFF679.1EA 0 GBP
CHMPCFF679.1 FLUO005512-1G FLUO005512-5G
5-(3-Pyridyl)-1H-tetrazole
5-(3-Pyridyl)-1H-tetrazole
Formula: C₆H₅N₅
MW: 147.14 g/mol
MDL Number: MFCD00040281
CAS Number: 3250-74-6
EINECS: 426-810-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR