5-(2-Pyridyl)-1H-tetrazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(1H-Tetrazol-5-yl)pyridine, 5-(2-Pyridyl)-1H-tetrazole

005511-1G 005511-5G
FLUO005511-1GEA 0 GBP
FLUO005511-1G FLUO005511-5G
5-(2-Pyridyl)-1H-tetrazole
5-(2-Pyridyl)-1H-tetrazole
Formula: C₆H₅N₅
MW: 147.14 g/mol
MDL Number: MFCD00068114
CAS Number: 33893-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR