4-(Tributylstannyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51168G1 AS51168G5
ACTIAS51168G1EA 140 GBP
ACTIAS51168G1 ACTIAS51168G5
4-(Tributylstannyl)thiazole
4-(Tributylstannyl)thiazole
Formula: C₁₅H₂₉NSSn
MW: 374.17 g/mol
MDL Number: MFCD01319073
CAS Number: 173979-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR