4,5-Dimethyl-2-(tributylstannyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51244G1 AS51244G5
ACTIAS51244G1EA 291 GBP
ACTIAS51244G1 ACTIAS51244G5
4,5-Dimethyl-2-(tributylstannyl)thiazole
4,5-Dimethyl-2-(tributylstannyl)thiazole
Formula: C₁₇H₃₃NSSn
MDL Number: MFCD09265485
CAS Number: 938181-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR