2-Methoxy-4-(tributylstannyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51171G1 AS51171G5
ACTIAS51171G1EA 148 GBP
ACTIAS51171G1 ACTIAS51171G5
2-Methoxy-4-(tributylstannyl)thiazole
2-Methoxy-4-(tributylstannyl)thiazole
Formula: C₁₆H₃₁NOSSn
MW: 404.20 g/mol
MDL Number: MFCD07787372
CAS Number: 927391-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR