2-(Tributylstannyl)pyrimidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

721174-1G 721174-5G
SIAL721174-1GEA 199 GBP
SIAL721174-1G SIAL721174-5G
2-(Tributylstannyl)pyrimidine, Sigma-Aldrich®
2-(Tributylstannyl)pyrimidine
Formula: C₁₆H₃₀N₂Sn
MDL Number: MFCD01319085
CAS Number: 153435-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR