2-Methoxy-6-(tributylstannyl)pyrazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS51202G1
AS51202G5
ACTIAS51202G1EA
214
GBP
ACTIAS51202G1
ACTIAS51202G5
2-Methoxy-6-(tributylstannyl)pyrazine
2-Methoxy-6-(tributylstannyl)pyrazine
Formula:
C₁₇H₃₂N₂OSn MW: 399.15 g/mol |
MDL Number:
MFCD08275735 CAS Number: 1105511-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...