2-Methoxy-3-(tributylstannyl)pyrazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51237G1 AS51237G5
ACTIAS51237G1EA 137 GBP
ACTIAS51237G1 ACTIAS51237G5
2-Methoxy-3-(tributylstannyl)pyrazine
2-Methoxy-3-(tributylstannyl)pyrazine
Formula: C₁₇H₃₂N₂OSn
MDL Number: MFCD09025812
CAS Number: 1105511-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR