2,6-Dichloro-3-(tributylstannyl)pyrazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51176G1 AS51176G5
ACTIAS51176G1EA 223 GBP
ACTIAS51176G1 ACTIAS51176G5
2,6-Dichloro-3-(tributylstannyl)pyrazine
2,6-Dichloro-3-(tributylstannyl)pyrazine
Formula: C₁₆H₂₈Cl₂N₂Sn
MDL Number: MFCD07787382
CAS Number: 446285-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR