1-Methyl-2-(tributylstannyl)imidazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51165G1 AS51165G5
ACTIAS51165G1EA 182 GBP
ACTIAS51165G1 ACTIAS51165G5
1-Methyl-2-(tributylstannyl)imidazole
1-Methyl-2-(tributylstannyl)imidazole
Formula: C₁₆H₃₂N₂Sn
MW: 371.15 g/mol
MDL Number: MFCD01319030
CAS Number: 105494-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR