Methyl-8-chloro-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6954-5G HC-6954-1G
COBBHC-6954-5GEA 0 GBP
COBBHC-6954-5G COBBHC-6954-1G
Methyl-8-chloro-4-hydroxyquinoline-2-carboxylate
Methyl 8-chloro-4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₈ClNO₃
MDL Number: MFCD11053822
CAS Number: 1065074-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR