2,6-Bis(2,2,2-trifluoroethoxy)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004992-1G
004992-5G
FLUO004992-1GEA
0
GBP
FLUO004992-1G
FLUO004992-5G
2,6-Bis(2,2,2-trifluoroethoxy)benzonitrile
2,6-Bis(2,2,2-trifluoroethoxy)benzonitrile
Formula:
C₁₁H₁₁F₆NO₂ |
MDL Number:
MFCD00109080 CAS Number: 93624-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...