VWR® Electrode Holders
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
RDBT002015EA 0 GBP
RDBT002015
VWR® Electrode Holders
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Plain borosilicate tubes for insertion of a wire electrode so that the electrode is insulated from the clamp or holder and protected from mechanical damage. The tube also helps the flexible wire electrode be positioned correctly in a rigid support.