SPE cartridges, Dionex™, SolEx™
Supplier: DIONEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
074410 074411 074412 074413 074415 074416 074417 074589 074590 074623 075895 075896 075897 075898
DION074410EA 0 GBP
DION074410 DION074411 DION074412 DION074413 DION074415 DION074416 DION074417 DION074589 DION074590 DION074623 DION075895 DION075896 DION075897 DION075898
SPE cartridges, Dionex™, SolEx™
Chromatography Columns
Dionex™ SolEx™ SPE cartridges are designed for use with Dionex AutoTrace 280 SPE system or other SPE instrumentation and vacuum manifolds. They are suitable for extracting organic pollutants from a liquid sample off-line using a solid-phase adsorbent.
  • Silica or carbon based absorbents are bound or encapsulated in a membrane filter, or packed in a cartridge
  • Phthalate-free version provides extraction of pesticides from water and perform trace analyses
  • Also used to extract contaminants from milk, fruit juices, urine and other matrices
Regular versions are available in 1, 3 and 6 ml barrels. C18 pthalate-free version is available in 6 ml barrels and feature teflon frits and are packed in mylar bags to minimize phthalate contamination.