3-Chloroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloropyridine-4-carboxylic acid

AS22746G25 AS22746G5
ACTIAS22746G25EA 73 GBP
ACTIAS22746G25 ACTIAS22746G5
3-Chloroisonicotinic acid
3-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD03094685
CAS Number: 88912-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR