4-Methyl-2-quinolinol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Lepidinol, 4-Methylquinolin-2-ol, 2-Hydroxy-4-methylquinoline
HC-2230-5G
HC-2230-1G
HC-2230-25G
COBBHC-2230-5GEA
0
GBP
COBBHC-2230-5G
COBBHC-2230-1G
COBBHC-2230-25G
4-Methyl-2-quinolinol
4-Methyl-2-quinolinol
Formula:
C₁₀H₉NO MW: 159.185 g/mol Boiling Pt: 346 °C (1013 hPa) Melting Pt: 220…222 °C |
MDL Number:
MFCD00006745 CAS Number: 607-66-9 EINECS: 210-139-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...