6-(4-Fluorophenyl)-3-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-(4-Fluorophenyl)pyridine-3-boronic acid

BB-8572-250M BB-8572-100M
COBBBB-8572-250MEA 0 GBP
COBBBB-8572-250M COBBBB-8572-100M
6-(4-Fluorophenyl)-3-pyridineboronic acid
6-(4-Fluorophenyl)-3-pyridineboronic acid
Formula: C₁₁H₉BFNO₂
MW: 217.01 g/mol
MDL Number: MFCD09037487
CAS Number: 1072944-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR