3-Methyl-4-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Picoline-4-boronic acid, (3-Methylpyridin-4-yl)boronic acid

BB-8406-1G BB-8406-250M
COBBBB-8406-1GEA 0 GBP
COBBBB-8406-1G COBBBB-8406-250M
3-Methyl-4-pyridineboronic acid
3-Methyl-4-pyridineboronic acid
Formula: C₆H₈BNO₂
MW: 136.9400 g/mol
MDL Number: MFCD03095365
CAS Number: 894808-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR