2-Propoxy-5-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (6-Propoxypyridin-3-yl)boronic acid

BB-5609-5G BB-5609-1G
COBBBB-5609-5GEA 0 GBP
COBBBB-5609-5G COBBBB-5609-1G
2-Propoxy-5-pyridineboronic acid
2-Propoxy-5-pyridineboronic acid
Formula: C₈H₁₂BNO₃
MW: 181.0000 g/mol
MDL Number: MFCD11855844
CAS Number: 1150114-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR