3-(4-Bromophenyl)-5-(1-chloro-2-methylpropan-2-yl)-1,2,4-oxadiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6435-5G HC-6435-1G
COBBHC-6435-5GEA 0 GBP
COBBHC-6435-5G COBBHC-6435-1G
3-(4-Bromophenyl)-5-(1-chloro-2-methylpropan-2-yl)-1,2,4-oxadiazole
3-(4-Bromophenyl)-5-(1-chloro-2-methylpropan-2-yl)-1,2,4-oxadiazole
Formula: C₁₂H₁₂BrClN₂O
MDL Number: MFCD10699657
CAS Number: 1033201-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR