Methyl-4-amino-3-methylbenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-amino-3-methylbenzoate

CA-4262-100G CA-4262-5G CA-4262-25G
COBBCA-4262-100GEA 0 GBP
COBBCA-4262-100G COBBCA-4262-5G COBBCA-4262-25G
Methyl-4-amino-3-methylbenzoate
Methyl 4-amino-3-methylbenzoate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00102230
CAS Number: 18595-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR