Methyl-2-amino-3-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-amino-3-nitrobenzoate

AS22118G1 AS22118G25 AS22118G5
ACTIAS22118G1EA 0 GBP
ACTIAS22118G1 ACTIAS22118G25 ACTIAS22118G5
Methyl-2-amino-3-nitrobenzoate
Methyl 2-amino-3-nitrobenzoate
Formula: C₈H₈N₂O₄
MW: 196.1600 g/mol
MDL Number: MFCD02093531
CAS Number: 57113-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR