Ethyl-2-amino-4-chlorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4498-5G CA-4498-25G CA-4498-1G
COBBCA-4498-5GEA 0 GBP
COBBCA-4498-5G COBBCA-4498-25G COBBCA-4498-1G
Ethyl-2-amino-4-chlorobenzoate
Ethyl 2-amino-4-chlorobenzoate
Formula: C₉H₁₀ClNO₂
MW: 199.630 g/mol
MDL Number: MFCD09878359
CAS Number: 60064-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR