3-Decylthiophene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SIAL456357-5GEA 350 GBP
SIAL456357-5G
3-Decylthiophene, Sigma-Aldrich®
3-Decylthiophene
Formula: C₁₄H₂₄S
MW: 224.40 g/mol
MDL Number: MFCD00143180
CAS Number: 65016-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR