3-Butylthiophene ≥98.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B1458-1G
B1458-5G
TCIAB1458-1GEA
0
GBP
TCIAB1458-1G
TCIAB1458-5G
3-Butylthiophene ≥98.0% (by GC)
3-Butylthiophene
Formula:
C₈H₁₂S MW: 140.2459 g/mol Boiling Pt: 183°C Flash Pt: 60°C |
MDL Number:
MFCD00143179 CAS Number: 34722-01-5 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...