3-Butylthiophene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

510424-1G 510424-5G
SIAL510424-1GEA 41.8 GBP
SIAL510424-1G SIAL510424-5G
3-Butylthiophene, Sigma-Aldrich®
3-Butylthiophene
Formula: C₈H₁₂S
MW: 140.2459 g/mol
MDL Number: MFCD00143179
CAS Number: 34722-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR