Methyl-5-bromo-2-hydroxyisonicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-bromo-2-hydroxyisonicotinate

CA-4141-5G CA-4141-1G
COBBCA-4141-5GEA 0 GBP
COBBCA-4141-5G COBBCA-4141-1G
Methyl-5-bromo-2-hydroxyisonicotinate
Methyl 5-bromo-2-hydroxyisonicotinate
Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
MDL Number: MFCD09027265
CAS Number: 913836-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR