Methyl-4-bromo-2,6-difluorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-bromo-2,6-difluorobenzoate

CA-4983-5G CA-4983-1G
COBBCA-4983-5GEA 0 GBP
COBBCA-4983-5G COBBCA-4983-1G
Methyl-4-bromo-2,6-difluorobenzoate
Methyl 4-bromo-2,6-difluorobenzoate
Formula: C₈H₅BrF₂O₂
MW: 251.0300 g/mol
MDL Number: MFCD06204169
CAS Number: 773134-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR