6,7-Dichloroquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7236-5G
PY-7236-1G
COBBPY-7236-5GEA
0
GBP
COBBPY-7236-5G
COBBPY-7236-1G
6,7-Dichloroquinoline
6,7-Dichloroquinoline
Formula:
C₉H₅Cl₂N |
MDL Number:
MFCD09878431 CAS Number: 40635-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...