2,6-Dichloroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1322-5G PY-1322-1G
COBBPY-1322-5GEA 0 GBP
COBBPY-1322-5G COBBPY-1322-1G
2,6-Dichloroquinoline
2,6-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.0500 g/mol
MDL Number: MFCD03427194
CAS Number: 1810-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR