(2,3-Difluoro-6-nitrophenyl)acetic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1354-5G AN-1354-1G
COBBAN-1354-5GEA 0 GBP
COBBAN-1354-5G COBBAN-1354-1G
(2,3-Difluoro-6-nitrophenyl)acetic acid
(2,3-Difluoro-6-nitrophenyl)acetic acid
Formula: C₈H₅F₂NO₄
MDL Number: MFCD11053839
CAS Number: 141428-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR