4-Cyano-2-fluorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-2-fluorobenzeneboronic acid pinacol ester

PN-8922-1G PN-8922-250M
COBBPN-8922-1GEA 0 GBP
COBBPN-8922-1G COBBPN-8922-250M
4-Cyano-2-fluorophenylboronic acid pinacol ester
4-Cyano-2-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
MW: 247.08 g/mol
MDL Number: MFCD09998162
CAS Number: 1035235-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR