2-Cyano-3-fluorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyano-3-fluorobenzeneboronic acid pinacol ester

PN-8822-1G PN-8822-5G
COBBPN-8822-1GEA 0 GBP
COBBPN-8822-1G COBBPN-8822-5G
2-Cyano-3-fluorophenylboronic acid pinacol ester
2-Cyano-3-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
MDL Number: MFCD09878541
CAS Number: 765916-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR