2,4,6-Trifluoroiodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021384-5G 021384-1G
FLUO021384-5GEA 0 GBP
FLUO021384-5G FLUO021384-1G
2,4,6-Trifluoroiodobenzene
2,4,6-Trifluoroiodobenzene
Formula: C₆H₂F₃I
MW: 257.98 g/mol
MDL Number: MFCD03095328
CAS Number: 41860-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR