2,3,4-Trifluoroiodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,2,3-Trifluoro-4-iodobenzene

021382-5G 021382-1G
FLUO021382-5GEA 0 GBP
FLUO021382-5G FLUO021382-1G
2,3,4-Trifluoroiodobenzene
2,3,4-Trifluoroiodobenzene
Formula: C₆H₂F₃I
MW: 257.9798 g/mol
MDL Number: MFCD03788551
CAS Number: 459424-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR