1,4-Diiodo-2-fluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Fluoro-1,4-diiodobenzene
009150-5G
009150-25G
FLUO009150-5GEA
0
GBP
FLUO009150-5G
FLUO009150-25G
1,4-Diiodo-2-fluorobenzene
1,4-Diiodo-2-fluorobenzene
Formula:
C₆H₃FI₂ MW: 347.9000 g/mol |
MDL Number:
MFCD00061107 CAS Number: 147808-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...