4-Bromo-5-fluoro-2-methylbenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4642-1G CA-4642-5G
COBBCA-4642-1GEA 0 GBP
COBBCA-4642-1G COBBCA-4642-5G
4-Bromo-5-fluoro-2-methylbenzonitrile
4-Bromo-5-fluoro-2-methylbenzonitrile
Formula: C₈H₅BrFN
MW: 214.0344 g/mol
MDL Number: MFCD07782074
CAS Number: 916792-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR